Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CATGACGCAGTGTCTCCCAGGTGAG[C/T]CTGCCGCCCTCCCTCGGCGGGGCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603842 MIM: 610057 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
NDUFB7 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
NDUFB7 - NADH:ubiquinone oxidoreductase subunit B7 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004146.5 | Intron | NP_004137.2 | ||||
XM_011528039.2 | Intron | XP_011526341.1 |
TECR - trans-2,3-enoyl-CoA reductase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001321170.1 | Intron | NP_001308099.1 | ||||
NM_138501.5 | Intron | NP_612510.1 | ||||
XM_006722945.1 | Intron | XP_006723008.1 | ||||
XM_011528442.2 | Intron | XP_011526744.1 |
Set Membership: |
HapMap |