Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGAATCAAAGTCTCCTGTCTTTG[T/C]CTGCTGATGCTCATGGAGATTTACA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608452 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
PDGFC PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
PDGFC - platelet derived growth factor C | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_016205.2 | Intron | NP_057289.1 | ||||
XM_011532124.2 | Intron | XP_011530426.1 | ||||
XM_017008455.1 | Intron | XP_016863944.1 | ||||
XM_017008456.1 | Intron | XP_016863945.1 |