Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TAACAAATCTGGATCTCTTGCCCAA[A/T]GGGTGACGTTGTGCTGAAGGAGGAG
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608166 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SEMA3E PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SEMA3E - semaphorin 3E | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001178129.1 | 5271 | UTR 3 | NP_001171600.1 | |||
NM_012431.2 | 5271 | UTR 3 | NP_036563.1 |
Set Membership: |
HapMap |