Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
14 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605743 MIM: 603305 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
HHAT PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
HHAT - hedgehog acyltransferase | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
KCNH1 - potassium voltage-gated channel subfamily H member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_002238.3 | 4302 | UTR 3 | NP_002229.1 | |||
NM_172362.2 | 4302 | UTR 3 | NP_758872.1 | |||
XM_017001246.1 | 4302 | UTR 3 | XP_016856735.1 |
Set Membership: |
HapMap |