Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616823 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
DOPEY1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
DOPEY1 - dopey family member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001199942.1 | Intron | NP_001186871.1 | ||||
NM_015018.3 | Intron | NP_055833.2 | ||||
XM_011535619.2 | Intron | XP_011533921.1 | ||||
XM_017010559.1 | Intron | XP_016866048.1 | ||||
XM_017010560.1 | Intron | XP_016866049.1 | ||||
XM_017010561.1 | Intron | XP_016866050.1 | ||||
XM_017010562.1 | Intron | XP_016866051.1 | ||||
XM_017010563.1 | Intron | XP_016866052.1 | ||||
XM_017010564.1 | Intron | XP_016866053.1 | ||||
XM_017010565.1 | Intron | XP_016866054.1 | ||||
XM_017010566.1 | Intron | XP_016866055.1 | ||||
XM_017010567.1 | Intron | XP_016866056.1 | ||||
XM_017010568.1 | Intron | XP_016866057.1 | ||||
XM_017010569.1 | Intron | XP_016866058.1 | ||||
XM_017010570.1 | Intron | XP_016866059.1 | ||||
XM_017010571.1 | Intron | XP_016866060.1 | ||||
XM_017010572.1 | Intron | XP_016866061.1 | ||||
XM_017010573.1 | Intron | XP_016866062.1 | ||||
XM_017010574.1 | Intron | XP_016866063.1 |