Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CACAGAGGGTGAAGAAGCCAGCCAC[A/C]ACCATCATGATGACTGATATGGCCA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606912 MIM: 613472 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MIR6882 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
MIR6882 - microRNA 6882 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SCAMP2 - secretory carrier membrane protein 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001320778.1 | 1054 | Silent Mutation | GTG,GTT | V,V 310 | NP_001307707.1 | |
NM_005697.4 | 1054 | Silent Mutation | GTG,GTT | V,V 267 | NP_005688.2 | |
XM_017021856.1 | 1054 | Silent Mutation | GTG,GTT | V,V 211 | XP_016877345.1 |
ULK3 - unc-51 like kinase 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |