Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 605994 MIM: 613725 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CYP39A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CYP39A1 - cytochrome P450 family 39 subfamily A member 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC25A27 - solute carrier family 25 member 27 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001204051.1 | Intron | NP_001190980.1 | ||||
NM_001204052.1 | Intron | NP_001190981.1 | ||||
NM_004277.4 | Intron | NP_004268.3 | ||||
XM_005249483.2 | Intron | XP_005249540.1 | ||||
XM_017011512.1 | Intron | XP_016867001.1 |
Set Membership: |
HapMap |