Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610221 MIM: 609742 MIM: 616659 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AKT1S1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
AKT1S1 - AKT1 substrate 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
IL4I1 - interleukin 4 induced 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MIR4750 - microRNA 4750 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TBC1D17 - TBC1 domain family member 17 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001168222.1 | 451 | Missense Mutation | GAC,GGC | D,G 51 | NP_001161694.1 | |
NM_024682.2 | 451 | Missense Mutation | GAC,GGC | D,G 84 | NP_078958.2 | |
XM_011527317.2 | 451 | Missense Mutation | GAC,GGC | D,G 84 | XP_011525619.1 |
Set Membership: |
HapMap |