Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AGTGGGGGAGGTTTCAAACCATAGA[A/C]CCAGTCTAATTTGATTATGCACAGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604494 MIM: 601203 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
IL18R1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
IL18R1 - interleukin 18 receptor 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001282399.1 | Intron | NP_001269328.1 | ||||
NM_003855.3 | Intron | NP_003846.1 | ||||
XM_005264039.3 | Intron | XP_005264096.1 | ||||
XM_005264040.3 | Intron | XP_005264097.1 | ||||
XM_011512099.1 | Intron | XP_011510401.1 | ||||
XM_017005181.1 | Intron | XP_016860670.1 | ||||
XM_017005182.1 | Intron | XP_016860671.1 | ||||
XM_017005183.1 | Intron | XP_016860672.1 | ||||
XM_017005184.1 | Intron | XP_016860673.1 |
IL1RL1 - interleukin 1 receptor like 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |