Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300490 MIM: 300588 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SH2D1A PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SH2D1A - SH2 domain containing 1A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TENM1 - teneurin transmembrane protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001163278.1 | 10285 | UTR 3 | NP_001156750.1 | |||
NM_001163279.1 | 10285 | UTR 3 | NP_001156751.1 | |||
NM_014253.3 | 10285 | UTR 3 | NP_055068.2 | |||
XM_011531230.2 | 10285 | UTR 3 | XP_011529532.1 | |||
XM_011531236.2 | 10285 | UTR 3 | XP_011529538.2 | |||
XM_011531237.2 | 10285 | UTR 3 | XP_011529539.1 | |||
XM_017029208.1 | 10285 | UTR 3 | XP_016884697.1 | |||
XM_017029209.1 | 10285 | UTR 3 | XP_016884698.1 | |||
XM_017029210.1 | 10285 | UTR 3 | XP_016884699.1 | |||
XM_017029211.1 | 10285 | UTR 3 | XP_016884700.1 | |||
XM_017029212.1 | 10285 | UTR 3 | XP_016884701.1 | |||
XM_017029213.1 | 10285 | UTR 3 | XP_016884702.1 | |||
XM_017029214.1 | 10285 | UTR 3 | XP_016884703.1 | |||
XM_017029215.1 | 10285 | Intron | XP_016884704.1 | |||
XM_017029216.1 | 10285 | Intron | XP_016884705.1 |