Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACACTTTTAGTCCTAAGCCTTACAC[A/G]TAAGACATACTCAATCTGTATGAAC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 114890 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CEACAM5 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CEACAM5 - carcinoembryonic antigen related cell adhesion molecule 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001291484.2 | Intron | NP_001278413.1 | ||||
NM_001308398.1 | Intron | NP_001295327.1 | ||||
NM_004363.5 | Intron | NP_004354.3 | ||||
XM_005258413.3 | Intron | XP_005258470.2 | ||||
XM_011526322.1 | Intron | XP_011524624.1 | ||||
XM_017026145.1 | Intron | XP_016881634.1 | ||||
XM_017026146.1 | Intron | XP_016881635.1 |