Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
CTAAAGAATATATTTTTGAACTCCT[C/T]GTTTCTGCGCTGTCCTTCTTAGCCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 114212 MIM: 612638 MIM: 603327 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CAPS PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CAPS - calcyphosine | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NDUFA11 - NADH:ubiquinone oxidoreductase subunit A11 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
RANBP3 - RAN binding protein 3 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
VMAC - vimentin-type intermediate filament associated coiled-coil protein | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001017921.3 | 1278 | UTR 3 | NP_001017921.1 |