Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
20 submissions
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 608353 MIM: 616755 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
AZIN2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
TRIM62 - tripartite motif containing 62 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_018207.2 | 3494 | UTR 3 | NP_060677.2 | |||
XM_011541705.2 | 3494 | UTR 3 | XP_011540007.1 | |||
XM_017001629.1 | 3494 | UTR 3 | XP_016857118.1 | |||
XM_017001630.1 | 3494 | UTR 3 | XP_016857119.1 | |||
XM_017001631.1 | 3494 | Intron | XP_016857120.1 |
Set Membership: |
HapMap |