Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608188 MIM: 608187 | ||||||||||||||||||||
Literature Links: |
CRLS1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CRLS1 - cardiolipin synthase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001127458.1 | Intron | NP_001120930.1 | ||||
NM_001323561.1 | Intron | NP_001310490.1 | ||||
NM_001323562.1 | Intron | NP_001310491.1 | ||||
NM_001323563.1 | Intron | NP_001310492.1 | ||||
NM_001323564.1 | Intron | NP_001310493.1 | ||||
NM_019095.5 | Intron | NP_061968.1 | ||||
XM_005260738.4 | Intron | XP_005260795.1 | ||||
XM_011529263.1 | Intron | XP_011527565.1 |
MCM8 - minichromosome maintenance 8 homologous recombination repair factor | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
MCM8-AS1 - MCM8 antisense RNA 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |