Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GAATGTACTGGGTGGTGTCCATCTG[G/T]TCTTGGATGGTGTAAGCATAAGGAT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616174 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CKAP2L PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CKAP2L - cytoskeleton associated protein 2 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304361.1 | Intron | NP_001291290.1 | ||||
NM_152515.4 | Intron | NP_689728.3 | ||||
XM_011510666.2 | Intron | XP_011508968.1 |
NT5DC4 - 5'-nucleotidase domain containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_011512262.2 | Intron | XP_011510564.1 | ||||
XM_011512265.2 | Intron | XP_011510567.1 | ||||
XM_017005474.1 | Intron | XP_016860963.1 | ||||
XM_017005475.1 | Intron | XP_016860964.1 | ||||
XM_017005476.1 | Intron | XP_016860965.1 | ||||
XM_017005477.1 | Intron | XP_016860966.1 | ||||
XM_017005478.1 | Intron | XP_016860967.1 |
Set Membership: |
HapMap |