Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604151 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
UBE2E3 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
UBE2E3 - ubiquitin conjugating enzyme E2 E3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278554.1 | 1774 | Intron | NP_001265483.1 | |||
NM_001278555.1 | 1774 | Intron | NP_001265484.1 | |||
NM_006357.3 | 1774 | Intron | NP_006348.1 | |||
NM_182678.2 | 1774 | Intron | NP_872619.1 | |||
XM_005246244.2 | 1774 | Intron | XP_005246301.1 | |||
XM_011510492.2 | 1774 | UTR 3 | XP_011508794.1 | |||
XM_017003166.1 | 1774 | Intron | XP_016858655.1 | |||
XM_017003167.1 | 1774 | Intron | XP_016858656.1 |
Set Membership: |
HapMap |