Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GGGTTGTTGTATAGGGACCTAGATA[C/T]GCACACTTCAGAAGGGTCCTGGGAA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 611692 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
ZBTB34 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
ZBTB34 - zinc finger and BTB domain containing 34 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001099270.1 | Intron | NP_001092740.1 | ||||
XM_005251989.4 | Intron | XP_005252046.1 | ||||
XM_011518699.2 | Intron | XP_011517001.1 |