Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 180220 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RARB PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
RARB - retinoic acid receptor beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000965.4 | Intron | NP_000956.2 | ||||
NM_001290216.2 | Intron | NP_001277145.1 | ||||
NM_001290217.1 | Intron | NP_001277146.1 | ||||
NM_001290266.1 | Intron | NP_001277195.1 | ||||
NM_001290276.1 | Intron | NP_001277205.1 | ||||
NM_001290277.1 | Intron | NP_001277206.1 | ||||
NM_001290300.1 | Intron | NP_001277229.1 | ||||
NM_016152.3 | Intron | NP_057236.1 |
Set Membership: |
HapMap |