Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GCTGAAAGGTTGCTTTAGATTTTCC[A/G]GAGAAAGACTATAAATCCTTAAATT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CDADC1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CDADC1 - cytidine and dCMP deaminase domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001193478.1 | Intron | NP_001180407.1 | ||||
NM_030911.3 | Intron | NP_112173.1 | ||||
XM_006719871.3 | Intron | XP_006719934.1 | ||||
XM_011535250.2 | Intron | XP_011533552.1 | ||||
XM_011535251.1 | Intron | XP_011533553.1 | ||||
XM_011535253.1 | Intron | XP_011533555.1 | ||||
XM_017020778.1 | Intron | XP_016876267.1 | ||||
XM_017020779.1 | Intron | XP_016876268.1 | ||||
XM_017020780.1 | Intron | XP_016876269.1 | ||||
XM_017020781.1 | Intron | XP_016876270.1 | ||||
XM_017020782.1 | Intron | XP_016876271.1 | ||||
XM_017020783.1 | Intron | XP_016876272.1 | ||||
XM_017020784.1 | Intron | XP_016876273.1 |
LOC105370203 - uncharacterized LOC105370203 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |