Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609085 MIM: 610930 MIM: 611052 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
FBXL19 PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
FBXL19 - F-box and leucine rich repeat protein 19 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001099784.2 | 103 | Intron | NP_001093254.2 | |||
NM_001282351.1 | 103 | Intron | NP_001269280.1 |
ORAI3 - ORAI calcium release-activated calcium modulator 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_152288.2 | 103 | UTR 5 | NP_689501.1 |
SETD1A - SET domain containing 1A | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |