Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
20 submissions
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 179520 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RAP1A PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
RAP1A - RAP1A, member of RAS oncogene family | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001010935.2 | Intron | NP_001010935.1 | ||||
NM_001291896.1 | Intron | NP_001278825.1 | ||||
NM_002884.3 | Intron | NP_002875.1 | ||||
XM_017001960.1 | Intron | XP_016857449.1 | ||||
XM_017001961.1 | Intron | XP_016857450.1 | ||||
XM_017001962.1 | Intron | XP_016857451.1 | ||||
XM_017001963.1 | Intron | XP_016857452.1 | ||||
XM_017001964.1 | Intron | XP_016857453.1 |
TMIGD3 - transmembrane and immunoglobulin domain containing 3 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001081976.2 | Intron | NP_001075445.1 | ||||
NM_001302680.1 | Intron | NP_001289609.1 | ||||
NM_020683.6 | Intron | NP_065734.5 |
Set Membership: |
HapMap |