Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATTTTTCTCTAAGGGGCACGTGATA[C/T]ACAAGAGCTTGTGGTGGGTAGCTCA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
41 submissions
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606373 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
FMN2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
FMN2 - formin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001305424.1 | Intron | NP_001292353.1 | ||||
NM_020066.4 | Intron | NP_064450.3 | ||||
XM_011544237.2 | Intron | XP_011542539.1 | ||||
XM_017001837.1 | Intron | XP_016857326.1 | ||||
XM_017001838.1 | Intron | XP_016857327.1 | ||||
XM_017001839.1 | Intron | XP_016857328.1 | ||||
XM_017001840.1 | Intron | XP_016857329.1 | ||||
XM_017001841.1 | Intron | XP_016857330.1 | ||||
XM_017001842.1 | Intron | XP_016857331.1 | ||||
XM_017001843.1 | Intron | XP_016857332.1 |
RPS7P5 - ribosomal protein S7 pseudogene 5 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |