Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
|||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
ARMC2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
ARMC2 - armadillo repeat containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001286609.1 | Intron | NP_001273538.1 | ||||
NM_032131.5 | Intron | NP_115507.4 | ||||
XM_005267154.3 | Intron | XP_005267211.1 | ||||
XM_005267155.2 | Intron | XP_005267212.1 | ||||
XM_005267157.3 | Intron | XP_005267214.1 | ||||
XM_006715573.2 | Intron | XP_006715636.1 | ||||
XM_006715574.2 | Intron | XP_006715637.1 | ||||
XM_011536166.1 | Intron | XP_011534468.1 | ||||
XM_011536167.1 | Intron | XP_011534469.1 | ||||
XM_011536168.2 | Intron | XP_011534470.1 | ||||
XM_011536170.2 | Intron | XP_011534472.1 | ||||
XM_011536172.2 | Intron | XP_011534474.1 | ||||
XM_011536176.2 | Intron | XP_011534478.1 | ||||
XM_017011350.1 | Intron | XP_016866839.1 | ||||
XM_017011351.1 | Intron | XP_016866840.1 | ||||
XM_017011352.1 | Intron | XP_016866841.1 |