Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 300303 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
RPS6KA6 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
RPS6KA6 - ribosomal protein S6 kinase A6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014496.4 | Intron | NP_055311.1 | ||||
XM_011530917.2 | Intron | XP_011529219.1 | ||||
XM_011530918.2 | Intron | XP_011529220.1 | ||||
XM_011530919.2 | Intron | XP_011529221.1 | ||||
XM_011530920.2 | Intron | XP_011529222.1 | ||||
XM_017029423.1 | Intron | XP_016884912.1 | ||||
XM_017029424.1 | Intron | XP_016884913.1 | ||||
XM_017029425.1 | Intron | XP_016884914.1 | ||||
XM_017029426.1 | Intron | XP_016884915.1 |
Set Membership: |
HapMap |