Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
MIM: 610298 MIM: 617015 | |||||||||||||||||||||||
Literature Links: |
PHLDB2 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
PHLDB2 - pleckstrin homology like domain family B member 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001134437.1 | Intron | NP_001127909.1 | ||||
NM_001134438.1 | Intron | NP_001127910.1 | ||||
NM_001134439.1 | Intron | NP_001127911.1 | ||||
NM_145753.2 | Intron | NP_665696.1 |
PLCXD2 - phosphatidylinositol specific phospholipase C X domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001185106.1 | Intron | NP_001172035.1 | ||||
NM_153268.3 | Intron | NP_695000.1 |