Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 603672 | ||||||||||||||||||||
Literature Links: |
CBFA2T2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CBFA2T2 - CBFA2/RUNX1 translocation partner 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001032999.2 | Intron | NP_001028171.1 | ||||
NM_001039709.1 | Intron | NP_001034798.1 | ||||
NM_005093.3 | Intron | NP_005084.1 | ||||
XM_011529101.2 | Intron | XP_011527403.1 | ||||
XM_011529102.2 | Intron | XP_011527404.1 | ||||
XM_011529103.2 | Intron | XP_011527405.1 | ||||
XM_011529107.2 | Intron | XP_011527409.1 | ||||
XM_017028121.1 | Intron | XP_016883610.1 | ||||
XM_017028122.1 | Intron | XP_016883611.1 | ||||
XM_017028123.1 | Intron | XP_016883612.1 | ||||
XM_017028124.1 | Intron | XP_016883613.1 | ||||
XM_017028125.1 | Intron | XP_016883614.1 |