Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613593 MIM: 613594 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
BTN3A1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
BTN3A1 - butyrophilin subfamily 3 member A1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001145008.1 | Intron | NP_001138480.1 | ||||
NM_001145009.1 | Intron | NP_001138481.1 | ||||
NM_007048.5 | Intron | NP_008979.3 | ||||
NM_194441.2 | Intron | NP_919423.1 | ||||
XM_005248833.2 | Intron | XP_005248890.1 | ||||
XM_005248834.3 | Intron | XP_005248891.1 | ||||
XM_006714983.2 | Intron | XP_006715046.1 |
Set Membership: |
HapMap |