Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616880 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TBC1D22B PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
TBC1D22B - TBC1 domain family member 22B | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_017772.3 | Intron | NP_060242.2 | ||||
XM_011514738.2 | Intron | XP_011513040.1 | ||||
XM_011514739.1 | Intron | XP_011513041.1 |
TMEM217 - transmembrane protein 217 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001162900.1 | Intron | NP_001156372.1 | ||||
NM_001286401.1 | Intron | NP_001273330.1 | ||||
NM_145316.3 | Intron | NP_660359.2 | ||||
XM_011514362.1 | Intron | XP_011512664.1 | ||||
XM_011514363.2 | Intron | XP_011512665.1 | ||||
XM_011514364.2 | Intron | XP_011512666.1 | ||||
XM_011514365.2 | Intron | XP_011512667.1 | ||||
XM_011514366.2 | Intron | XP_011512668.1 | ||||
XM_011514367.2 | Intron | XP_011512669.1 | ||||
XM_011514369.2 | Intron | XP_011512671.1 | ||||
XM_011514370.2 | Intron | XP_011512672.1 |
Set Membership: |
HapMap |