Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 606914 MIM: 614548 | |||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PKIB PubMed Links | |||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PKIB - protein kinase (cAMP-dependent, catalytic) inhibitor beta | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001270393.1 | Intron | NP_001257322.1 | ||||
NM_001270394.1 | Intron | NP_001257323.1 | ||||
NM_001270395.1 | Intron | NP_001257324.1 | ||||
NM_032471.5 | Intron | NP_115860.1 | ||||
NM_181794.2 | Intron | NP_861459.1 | ||||
NM_181795.2 | Intron | NP_861460.1 | ||||
XM_011535930.2 | Intron | XP_011534232.1 | ||||
XM_011535931.2 | Intron | XP_011534233.1 | ||||
XM_011535932.2 | Intron | XP_011534234.1 | ||||
XM_011535933.2 | Intron | XP_011534235.1 | ||||
XM_011535934.2 | Intron | XP_011534236.1 | ||||
XM_011535935.2 | Intron | XP_011534237.1 | ||||
XM_011535936.1 | Intron | XP_011534238.1 | ||||
XM_011535937.2 | Intron | XP_011534239.1 |
SERINC1 - serine incorporator 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |