Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||
Phenotype: |
MIM: 608200 | ||||||||||||||||||||
Literature Links: |
CDK5RAP1 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
CDK5RAP1 - CDK5 regulatory subunit associated protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001278167.1 | Intron | NP_001265096.1 | ||||
NM_001278168.1 | Intron | NP_001265097.1 | ||||
NM_001278169.1 | Intron | NP_001265098.1 | ||||
NM_016082.4 | Intron | NP_057166.4 | ||||
NM_016408.3 | Intron | NP_057492.2 | ||||
XM_011528856.2 | Intron | XP_011527158.1 | ||||
XM_011528857.2 | Intron | XP_011527159.1 | ||||
XM_011528858.1 | Intron | XP_011527160.1 | ||||
XM_011528859.2 | Intron | XP_011527161.1 | ||||
XM_011528860.1 | Intron | XP_011527162.1 | ||||
XM_017027876.1 | Intron | XP_016883365.1 | ||||
XM_017027877.1 | Intron | XP_016883366.1 |