Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AATTTTCATATTTATAATAGCACTT[A/G]ACTCTTATAAAAGGCTTTGTACTTT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 600267 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PTPN13 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PTPN13 - protein tyrosine phosphatase, non-receptor type 13 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006264.2 | Intron | NP_006255.1 | ||||
NM_080683.2 | Intron | NP_542414.1 | ||||
NM_080684.2 | Intron | NP_542415.1 | ||||
NM_080685.2 | Intron | NP_542416.1 | ||||
XM_011532165.1 | Intron | XP_011530467.1 | ||||
XM_017008511.1 | Intron | XP_016864000.1 | ||||
XM_017008512.1 | Intron | XP_016864001.1 | ||||
XM_017008513.1 | Intron | XP_016864002.1 | ||||
XM_017008514.1 | Intron | XP_016864003.1 | ||||
XM_017008515.1 | Intron | XP_016864004.1 | ||||
XM_017008516.1 | Intron | XP_016864005.1 |
Set Membership: |
HapMap |