Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GACCTGTTCACCCCCTTTTCAGTTT[C/T]TCCGTTTGCATCTGAGGGATTCTTG
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 194363 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TMEM167A PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
TMEM167A - transmembrane protein 167A | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_174909.4 | Intron | NP_777569.1 |
XRCC4 - X-ray repair cross complementing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001318012.1 | Intron | NP_001304941.1 | ||||
NM_001318013.1 | Intron | NP_001304942.1 | ||||
NM_003401.4 | Intron | NP_003392.1 | ||||
NM_022406.3 | Intron | NP_071801.1 | ||||
NM_022550.3 | Intron | NP_072044.1 | ||||
XM_011543626.1 | Intron | XP_011541928.1 | ||||
XM_017009827.1 | Intron | XP_016865316.1 | ||||
XM_017009828.1 | Intron | XP_016865317.1 | ||||
XM_017009829.1 | Intron | XP_016865318.1 |
Set Membership: |
HapMap |