Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ATGAATTATATAAACATTACAAGGA[A/G]TTTACCACTGAGTTCATAATTATGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 164860 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MET PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
MET - MET proto-oncogene, receptor tyrosine kinase | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000245.3 | Intron | NP_000236.2 | ||||
NM_001127500.2 | Intron | NP_001120972.1 | ||||
NM_001324401.1 | Intron | NP_001311330.1 | ||||
NM_001324402.1 | Intron | NP_001311331.1 | ||||
XM_006715990.2 | Intron | XP_006716053.1 | ||||
XM_011516223.1 | Intron | XP_011514525.1 |