Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 604026 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GOSR1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GOSR1 - golgi SNAP receptor complex member 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001007024.1 | Intron | NP_001007025.1 | ||||
NM_001007025.1 | Intron | NP_001007026.1 | ||||
NM_004871.2 | Intron | NP_004862.1 | ||||
XM_005258070.4 | Intron | XP_005258127.1 | ||||
XM_005258071.4 | Intron | XP_005258128.1 | ||||
XM_005258072.2 | Intron | XP_005258129.1 | ||||
XM_017025374.1 | Intron | XP_016880863.1 | ||||
XM_017025375.1 | Intron | XP_016880864.1 | ||||
XM_017025376.1 | Intron | XP_016880865.1 | ||||
XM_017025377.1 | Intron | XP_016880866.1 |
Set Membership: |
HapMap |