Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACGGAGGAAGGTATCAGGGTAGTGG[A/C]CCTCTGGAGGAGGCAGGCTGGTGGA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602488 MIM: 603953 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CYTH2 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
CYTH2 - cytohesin 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_004228.6 | Intron | NP_004219.3 | ||||
NM_017457.5 | Intron | NP_059431.1 | ||||
XM_006723472.2 | Intron | XP_006723535.1 | ||||
XM_006723473.2 | Intron | XP_006723536.1 |
KCNJ14 - potassium voltage-gated channel subfamily J member 14 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
LOC105372430 - uncharacterized LOC105372430 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
Set Membership: |
HapMap |