Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACAGGCCCAGGCCATCCAGATCACT[A/G]TGAACCTTATCAGAAACTTCAGTCC
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 614551 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SERINC5 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese)
|
||||||
EUR
|
||||||||
AMR
|
SERINC5 - serine incorporator 5 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001174071.2 | Intron | NP_001167542.1 | ||||
NM_001174072.2 | Intron | NP_001167543.1 | ||||
NM_178276.6 | Intron | NP_840060.1 | ||||
XM_011543304.1 | Intron | XP_011541606.1 | ||||
XM_017009323.1 | Intron | XP_016864812.1 | ||||
XM_017009324.1 | Intron | XP_016864813.1 | ||||
XM_017009325.1 | Intron | XP_016864814.1 | ||||
XM_017009326.1 | Intron | XP_016864815.1 |
Set Membership: |
HapMap |