Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||
Literature Links: |
GPR160 PubMed Links | |||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR - Not Available | |||||
AMR - Not Available |
GPR160 - G protein-coupled receptor 160 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_014373.2 | Intron | NP_055188.1 | ||||
XM_005247346.3 | Intron | XP_005247403.1 | ||||
XM_005247347.3 | Intron | XP_005247404.1 | ||||
XM_006713582.3 | Intron | XP_006713645.1 | ||||
XM_011512678.2 | Intron | XP_011510980.1 | ||||
XM_017006161.1 | Intron | XP_016861650.1 | ||||
XM_017006162.1 | Intron | XP_016861651.1 | ||||
XM_017006163.1 | Intron | XP_016861652.1 |