Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609362 MIM: 605641 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
PLA2G15 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
PLA2G15 - phospholipase A2 group XV | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
SLC7A6 - solute carrier family 7 member 6 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001076785.2 | Intron | NP_001070253.1 | ||||
NM_003983.5 | Intron | NP_003974.3 | ||||
XM_011523433.1 | Intron | XP_011521735.1 | ||||
XM_011523434.1 | Intron | XP_011521736.1 | ||||
XM_011523438.1 | Intron | XP_011521740.1 | ||||
XM_011523439.1 | Intron | XP_011521741.1 | ||||
XM_017023851.1 | Intron | XP_016879340.1 |
Set Membership: |
HapMap |