Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTCACTCATGTCCCTATTTTAGGA[A/T]CTAACTTCACATCTAAGCTAGCATA
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 609431 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
MBIP PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
MBIP - MAP3K12 binding inhibitory protein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001144891.1 | Intron | NP_001138363.1 | ||||
NM_001308110.1 | Intron | NP_001295039.1 | ||||
NM_016586.2 | Intron | NP_057670.2 | ||||
XM_005267754.3 | Intron | XP_005267811.1 | ||||
XM_005267756.4 | Intron | XP_005267813.1 | ||||
XM_017021367.1 | Intron | XP_016876856.1 |