Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
1 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 614695 | ||||||||||||||||||||
Literature Links: |
RPRD2 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RPRD2 - regulation of nuclear pre-mRNA domain containing 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001297673.1 | Intron | NP_001284602.1 | ||||
NM_001297674.1 | Intron | NP_001284603.1 | ||||
NM_015203.4 | Intron | NP_056018.2 | ||||
XM_005245033.2 | Intron | XP_005245090.1 | ||||
XM_006711240.2 | Intron | XP_006711303.1 | ||||
XM_011509344.2 | Intron | XP_011507646.1 | ||||
XM_017000763.1 | Intron | XP_016856252.1 | ||||
XM_017000764.1 | Intron | XP_016856253.1 | ||||
XM_017000765.1 | Intron | XP_016856254.1 | ||||
XM_017000766.1 | Intron | XP_016856255.1 | ||||
XM_017000767.1 | Intron | XP_016856256.1 |