Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 602461 | ||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
SIRPA PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese)
|
||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
SIRPA - signal regulatory protein alpha | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001040022.1 | Intron | NP_001035111.1 | ||||
NM_001040023.1 | Intron | NP_001035112.1 | ||||
NM_080792.2 | Intron | NP_542970.1 | ||||
XM_005260669.3 | Intron | XP_005260726.1 | ||||
XM_005260670.3 | Intron | XP_005260727.1 | ||||
XM_006723545.3 | Intron | XP_006723608.1 | ||||
XM_011529173.2 | Intron | XP_011527475.1 |
Set Membership: |
HapMap |