Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
TCTATAATAAATAAGATAAACAAGC[A/G]GGAACTAATAGAAATTTAATGTTGT
Species: |
Human | ||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 616174 | ||||||||||||||||||||||||||||||||||||||
Literature Links: |
CKAP2L PubMed Links | ||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | |||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | |||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | |||
EUR
|
|||||
AMR
|
CKAP2L - cytoskeleton associated protein 2 like | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001304361.1 | 3272 | UTR 3 | NP_001291290.1 | |||
NM_152515.4 | 3272 | UTR 3 | NP_689728.3 | |||
XM_011510666.2 | 3272 | UTR 3 | XP_011508968.1 |
NT5DC4 - 5'-nucleotidase domain containing 4 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_011512262.2 | 3272 | Intron | XP_011510564.1 | |||
XM_011512265.2 | 3272 | Intron | XP_011510567.1 | |||
XM_017005474.1 | 3272 | Intron | XP_016860963.1 | |||
XM_017005475.1 | 3272 | Intron | XP_016860964.1 | |||
XM_017005476.1 | 3272 | Intron | XP_016860965.1 | |||
XM_017005477.1 | 3272 | Intron | XP_016860966.1 | |||
XM_017005478.1 | 3272 | Intron | XP_016860967.1 |