Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
ACATCAATCTCCTAACTGCCTAAGT[C/T]TCACAAGTGTATCACTGACTCTCTT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
YTHDC1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
YTHDC1 - YTH domain containing 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
XM_005265706.2 | Intron | XP_005265763.1 | ||||
XM_005265707.2 | Intron | XP_005265764.1 | ||||
XM_005265708.2 | Intron | XP_005265765.1 | ||||
XM_017008832.1 | Intron | XP_016864321.1 | ||||
XM_017008833.1 | Intron | XP_016864322.1 |