Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 613927 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
C2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba) - Not Available | ||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
C2 - complement component 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_000063.5 | Intron | NP_000054.2 | ||||
NM_001145903.2 | Intron | NP_001139375.1 | ||||
NM_001178063.2 | Intron | NP_001171534.1 | ||||
NM_001282457.1 | Intron | NP_001269386.1 | ||||
NM_001282458.1 | Intron | NP_001269387.1 | ||||
NM_001282459.1 | Intron | NP_001269388.1 |
ZBTB12 - zinc finger and BTB domain containing 12 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |