Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
GTTTAGATTTTGTAATTACATGAAC[A/G]TATGCTATTGTCAAAAACTCACTGA
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610781 MIM: 603171 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GMPR2 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
GMPR2 - guanosine monophosphate reductase 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
NEDD8 - neural precursor cell expressed, developmentally down-regulated 8 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_006156.2 | Intron | NP_006147.1 |
NEDD8-MDP1 - NEDD8-MDP1 readthrough | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |