Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
CGATATGGACCAGTTCATGCGCAAG[A/C]GGATCGAGGCGATCCGCGGGCAGAT
Species: |
Human | ||||||||||||||||||||
dbSNP Submissions: |
5 submissions
|
||||||||||||||||||||
Phenotype: |
MIM: 611193 MIM: 190220 | ||||||||||||||||||||
Literature Links: |
RRP15 PubMed Links | ||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap |
---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU) - Not Available |
EAS - Not Available | African American - Not Available | YRI (Yoruba) - Not Available |
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available |
AFR - Not Available | Japanese - Not Available | JPT (Japanese) - Not Available |
EUR - Not Available | ||
AMR - Not Available |
RRP15 - ribosomal RNA processing 15 homolog | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TGFB2 - transforming growth factor beta 2 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
TGFB2-AS1 - TGFB2 antisense RNA 1 (head to head) | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |