Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | ||||||||||||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
||||||||||||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 610660 MIM: 609771 | ||||||||||||||||||||||||||||||||||||||||||||||||||
Literature Links: |
GLYR1 PubMed Links | ||||||||||||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
|||||||||||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU)
|
||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | JPT (Japanese)
|
||||||
AFR
|
Japanese - Not Available | CHB (Han Chinese)
|
||||||
EUR
|
||||||||
AMR
|
GLYR1 - glyoxylate reductase 1 homolog | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
UBN1 - ubinuclein 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001079514.2 | Intron | NP_001072982.1 | ||||
NM_001288656.1 | Intron | NP_001275585.1 | ||||
XM_005255277.3 | Intron | XP_005255334.1 | ||||
XM_005255278.3 | Intron | XP_005255335.1 | ||||
XM_005255279.3 | Intron | XP_005255336.1 | ||||
XM_011522465.1 | Intron | XP_011520767.1 | ||||
XM_011522466.2 | Intron | XP_011520768.1 | ||||
XM_011522467.1 | Intron | XP_011520769.1 | ||||
XM_017023189.1 | Intron | XP_016878678.1 | ||||
XM_017023190.1 | Intron | XP_016878679.1 | ||||
XM_017023191.1 | Intron | XP_016878680.1 | ||||
XM_017023192.1 | Intron | XP_016878681.1 |
Set Membership: |
HapMap |