Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
AAACATAAGTTTGAATACAGAGTTG[A/C]CTTTTTAAAATCTGGGCTTTTCTTT
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 603125 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
TPST1 PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
TPST1 - tyrosylprotein sulfotransferase 1 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_003596.3 | Intron | NP_003587.1 | ||||
XM_005250642.2 | Intron | XP_005250699.1 | ||||
XM_011516634.2 | Intron | XP_011514936.1 | ||||
XM_011516635.2 | Intron | XP_011514937.1 | ||||
XM_017012724.1 | Intron | XP_016868213.1 | ||||
XM_017012725.1 | Intron | XP_016868214.1 | ||||
XM_017012726.1 | Intron | XP_016868215.1 | ||||
XM_017012727.1 | Intron | XP_016868216.1 |
Set Membership: |
HapMap |