Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||||||||||||||
Phenotype: |
MIM: 118955 MIM: 608625 MIM: 611753 | |||||||||||||||||||||||||||||||||||||||||
Literature Links: |
CLTC PubMed Links | |||||||||||||||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | ||||||
---|---|---|---|---|---|---|---|---|
Global
|
Caucasian - Not Available | CEPH (CEU) - Not Available | ||||||
EAS
|
African American - Not Available | YRI (Yoruba)
|
||||||
SAS
|
Chinese - Not Available | CHB (Han Chinese) - Not Available | ||||||
AFR
|
Japanese - Not Available | JPT (Japanese) - Not Available | ||||||
EUR
|
||||||||
AMR
|
CLTC - clathrin heavy chain | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |
PTRH2 - peptidyl-tRNA hydrolase 2 | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001015509.2 | Intron | NP_001015509.1 | ||||
NM_016077.4 | Intron | NP_057161.1 | ||||
XM_011524887.1 | Intron | XP_011523189.1 |
VMP1 - vacuole membrane protein 1 | ||||||
---|---|---|---|---|---|---|
There are no transcripts associated with this gene. |