Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Species: |
Human | |||||||||||||||||||||||||||||
dbSNP Submissions: |
NA
|
|||||||||||||||||||||||||||||
Phenotype: |
MIM: 604167 | |||||||||||||||||||||||||||||
Literature Links: |
CTCF PubMed Links | |||||||||||||||||||||||||||||
Allele Nomenclature: |
||||||||||||||||||||||||||||||
Minor Allele Frequency: |
1000Genome | Applied Biosystems® | HapMap | |||
---|---|---|---|---|---|
Global - Not Available | Caucasian - Not Available | CEPH (CEU)
|
|||
EAS - Not Available | African American - Not Available | YRI (Yoruba)
|
|||
SAS - Not Available | Chinese - Not Available | CHB (Han Chinese) - Not Available | |||
AFR - Not Available | Japanese - Not Available | JPT (Japanese)
|
|||
EUR - Not Available | |||||
AMR - Not Available |
CTCF - CCCTC-binding factor | ||||||
---|---|---|---|---|---|---|
Transcript Accession | SNP Location | SNP Type | Observed Codons | Observed Amino Acid | Protein ID | |
NM_001191022.1 | Intron | NP_001177951.1 | ||||
NM_006565.3 | Intron | NP_006556.1 | ||||
XM_005255775.3 | Intron | XP_005255832.1 | ||||
XM_017022868.1 | Intron | XP_016878357.1 | ||||
XM_017022869.1 | Intron | XP_016878358.1 |
Set Membership: |
HapMap |